Gene Transcripts

Gene Symbol
Itgam
Entrez Gene ID
25021
Gene Name
integrin, alpha M
Gene Aliases
Cd11b
Chromosome Location
Chr. 1: 199495307 - 199545738 on Build Rnor_6.0
UniGene ID
Rn.54465
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012711.1 NP_036843.1 6 600
XM_006230214.2 XP_006230276.1 7 720
GenBank mRNA AF268593.1 - 6 600

View Full Product Details