Gene Transcripts

Gene Symbol
Olr1593
Entrez Gene ID
289255
Gene Name
olfactory receptor 1593
Gene Aliases
ratchr13-90101548-90102495_ORF
Chromosome Location
Chr. 13: 92224971 - 92225918 on Build Rnor_6.0
UniGene ID
Rn.142335
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000085.1 NP_001000085.1 1 521

View Full Product Details