Gene Transcripts

Gene Symbol
Dcdc2
Entrez Gene ID
291130
Gene Name
doublecortin domain containing 2
Gene Aliases
Chromosome Location
Chr. 17: 41838201 - 42031265 on Build Rnor_6.0
UniGene ID
Rn.45430
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106110.2 NP_001099580.2 9 1280
GenBank mRNA FM100065.1 - 3 390

View Full Product Details