Gene Transcripts

Gene Symbol
Uba5
Entrez Gene ID
300968
Gene Name
ubiquitin-like modifier activating enzyme 5
Gene Aliases
Ube1dc1
Chromosome Location
Chr. 8: 112579455 - 112594192 on Build Rnor_6.0
UniGene ID
Rn.228670
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009669.1 NP_001009669.1 6 647
GenBank mRNA BC088757.1 - 6 647

View Full Product Details