Gene Transcripts

Gene Symbol
Gpr45
Entrez Gene ID
301372
Gene Name
G protein-coupled receptor 45
Gene Aliases
Chromosome Location
Chr. 9: 49837868 - 49841337 on Build Rnor_6.0
UniGene ID
Rn.147923
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106906.1 NP_001100376.1 1 627

View Full Product Details