Gene Transcripts

Gene Symbol
Gorab
Entrez Gene ID
304923
Gene Name
golgin, RAB6-interacting
Gene Aliases
Scyl1bp1
Chromosome Location
Chr. 13: 81682207 - 81698827 on Build Rnor_6.0
UniGene ID
Rn.45710
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100563.1 NP_001094033.1 4 720
XM_008769650.1 XP_008767872.1 3 1296
GenBank mRNA BC086370.1 - 2 199
BC160825.1 - 4 720

View Full Product Details