Gene Transcripts

Gene Symbol
Ubac1
Entrez Gene ID
362087
Gene Name
UBA domain containing 1
Gene Aliases
Ubadc1
Chromosome Location
Chr. 3: 3453396 - 3476242 on Build Rnor_6.0
UniGene ID
Rn.6512
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007742.1 NP_001007743.1 4 529
XM_006233689.1 XP_006233751.1 4 556
GenBank mRNA BC083603.1 - 4 529
FQ210767.1 - 4 415

View Full Product Details