Gene Transcripts

Gene Symbol
Zfp870
Entrez Gene ID
362845
Gene Name
zinc finger protein 870
Gene Aliases
Zfp709l2
Chromosome Location
Chr. 7: 15158673 - 15172579 on Build Rnor_6.0
UniGene ID
Rn.41593
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163062.1 NP_001156534.1 4 1298
XM_006241082.2 XP_006241144.1 4 1385

View Full Product Details