Gene Transcripts

Gene Symbol
Kcnq1
Entrez Gene ID
84020
Gene Name
potassium channel, voltage-gated KQT-like subfamily Q, member 1
Gene Aliases
Kvlqt1
Chromosome Location
Chr. 1: 216293087 - 216630339 on Build Rnor_6.0
UniGene ID
Rn.9779
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032073.1 NP_114462.1 6 912
GenBank mRNA AJ133685.1 - 6 912
AY043394.1 - 5 652
U92655.1 - 5 424

View Full Product Details