Gene Transcripts

Gene Symbol
Dnaja2
Entrez Gene ID
84026
Gene Name
DnaJ (Hsp40) homolog, subfamily A, member 2
Gene Aliases
Cpr3, Dj3, DjA2, Dnj3, Hirip4, Rdj2
Chromosome Location
Chr. 19: 22569999 - 22588143 on Build Rnor_6.0
UniGene ID
Rn.3904
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032079.2 NP_114468.2 5 520
GenBank mRNA BC087010.1 - 5 520
FQ231743.1 - 3 276
U95727.1 - 5 554

View Full Product Details