Gene Transcripts

Gene Symbol
Lin7a
Entrez Gene ID
85327
Gene Name
lin-7 homolog a (C. elegans)
Gene Aliases
Veli1
Chromosome Location
Chr. 7: 49429613 - 49627835 on Build Rnor_6.0
UniGene ID
Rn.31766
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053514.1 NP_445966.1 305
XM_006241305.2 XP_006241367.1 3 208
GenBank mRNA AF090134.1 - 305
AF090135.1 - 305

View Full Product Details