Gene Transcripts

Gene Symbol
Hadh
Entrez Gene ID
113965
Gene Name
hydroxyacyl-CoA dehydrogenase
Gene Aliases
Hadhsc
Chromosome Location
Chr. 2: 236353445 - 236395067 on Build Rnor_6.0
UniGene ID
Rn.92789
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057186.2 NP_476534.1 1, 2 161
GenBank mRNA AF095449.1 - 1, 2 161
FQ212781.1 - 1, 2 141

View Full Product Details