Gene Transcripts

Gene Symbol
Il12rb1
Entrez Gene ID
171333
Gene Name
interleukin 12 receptor, beta 1
Gene Aliases
Il12rb
Chromosome Location
Chr. 16: 20370722 - 20383572 on Build Rnor_6.0
UniGene ID
Rn.162266
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170604.1 NP_001164075.1 15, 16 1861
XM_006252821.2 XP_006252883.1 16, 17 1957
GenBank mRNA AF083328.1 - 2, 3 146

View Full Product Details