Gene Transcripts

Gene Symbol
Dmp1
Entrez Gene ID
25312
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
DENTMAT
Chromosome Location
Chr. 14: 6889851 - 6923986 on Build Rnor_6.0
UniGene ID
Rn.19340
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203493.3 NP_987089.2 4 819
XM_006250622.2 XP_006250684.1 9 3437
XM_008769994.1 XP_008768216.1 5 1135
XM_008769995.1 XP_008768217.1 8 1554
GenBank mRNA BC129082.1 - 4 819

View Full Product Details