Gene Transcripts

Gene Symbol
Dhx37
Entrez Gene ID
288647
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 37
Gene Aliases
Chromosome Location
Chr. 12: 36594058 - 36614165 on Build Rnor_6.0
UniGene ID
Rn.24493
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105926.2 NP_001099396.2 10 1386

View Full Product Details