Gene Transcripts

Gene Symbol
Glmn
Entrez Gene ID
289437
Gene Name
glomulin, FKBP associated protein
Gene Aliases
Chromosome Location
Chr. 14: 3204390 - 3247703 on Build Rnor_6.0
UniGene ID
Rn.44333
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105993.1 NP_001099463.1 5 463
XM_008769921.1 XP_008768143.1 4 1070
XM_008769922.1 XP_008768144.1 5 464
XM_008769923.1 XP_008768145.1 5 543
XM_008769924.1 XP_008768146.1 4 1070
XM_008769925.1 XP_008768147.1 3 1129
XM_008769926.1 XP_008768148.1 3 256
GenBank mRNA BC083633.1 - 4 403
BC100158.1 - 5 576
BC162027.1 - 5 439

View Full Product Details