Gene Transcripts

Gene Symbol
Spint5p
Entrez Gene ID
408232
Gene Name
serine peptidase inhibitor, Kunitz type 5
Gene Aliases
Tpil4
Chromosome Location
Chr. 3: 161178117 - 161179744 on Build Rnor_6.0
UniGene ID
Rn.198173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008881.1 NP_001008881.1 2 447
XM_006235609.1 XP_006235671.1 2 453

View Full Product Details