Gene Transcripts

Gene Symbol
Fscb
Entrez Gene ID
500659
Gene Name
fibrous sheath CABYR binding protein
Gene Aliases
Chromosome Location
Chr. 6: 86220260 - 86223052 on Build Rnor_6.0
UniGene ID
Rn.162868
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271107.1 NP_001258036.1 1 746
GenBank mRNA BC098054.1 - 1 138

View Full Product Details