Gene Transcripts

Gene Symbol
Rbm15
Entrez Gene ID
684233
Gene Name
RNA binding motif protein 15
Gene Aliases
Chromosome Location
Chr. 2: 210079678 - 210090200 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749380.3 XP_003749428.1 1 3174
XM_006233197.2 XP_006233259.1 1 3175

View Full Product Details