Gene Transcripts

Gene Symbol
FGF7
Entrez Gene ID
2252
Gene Name
fibroblast growth factor 7
Gene Aliases
HBGF-7, KGF
Chromosome Location
Chr.15: 49423178 - 49487326 on Build GRCh38
Uni Gene
Hs.567268
Interrogated Sequence Translated Protein
RefSeq NM_002009.3 NP_002000.1
GenBank mRNA AK313160.1 -
BC010956.1 -
CR542002.1 -
DB206350.1 -
ENST00000267843.0 -
M60828.1 -
S81661.1 -

View Full Product Details