Gene Transcripts

Gene Symbol
Itgb2
Entrez Gene ID
16414
Gene Name
integrin beta 2
Gene Aliases
2E6, AI528527, Cd18, LAD, LCAMB, Lfa1, MF17
Chromosome Location
Chr.10: 77530328 - 77565681 on Build GRCm38
Uni Gene
Mm.1137
Interrogated Sequence Translated Protein
RefSeq NM_008404.4 NP_032430.2
GenBank mRNA AK088435.1 -
AK136502.1 -
AK161236.1 -
AK169587.1 -
BC145644.1 -
M31039.1 -
X14951.1 -

View Full Product Details