Gene Transcripts

Gene Symbol
Fgf7
Entrez Gene ID
14178
Gene Name
fibroblast growth factor 7
Gene Aliases
Kgf
Chromosome Location
Chr.2: 126001108 - 126091185 on Build GRCm38
Uni Gene
Mm.330557
Interrogated Sequence Translated Protein
RefSeq NM_008008.4 NP_032034.1
GenBank mRNA AK033934.1 -
AK037172.1 -
AK086639.1 -
AK139776.1 -
BC052847.1 -
Z22703.1 -

View Full Product Details