Gene Transcripts

Gene Symbol
Nfkbib
Entrez Gene ID
18036
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, beta
Gene Aliases
I(Kappa)B(beta), I-kappa-B-beta, IKB-beta, IKappaBbeta, IkB, IkBb, NF-kappa-BIB, ikB-B
Chromosome Location
Chr.7: 28758251 - 28766639 on Build GRCm38
Uni Gene
Mm.220333
Interrogated Sequence Translated Protein
GenBank mRNA AK010218.1 -
AK038631.1 -
AK156731.1 -
AK182221.1 -
AK202557.1 -
AK213461.1 -
AY176058.1 -
AY176059.1 -
BC021938.1 -
BM899229.1 -
U19799.1 -

View Full Product Details