Gene Transcripts

Gene Symbol
Sprr2g
Entrez Gene ID
20761
Gene Name
small proline-rich protein 2G
Gene Aliases
Chromosome Location
Chr. 3: 92373915 - 92375229 on Build GRCm38
UniGene ID
Mm.57083
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003548.1 2 518
GenBank mRNA AY158991.1 - 2 517
BC130233.1 - 2 501

View Full Product Details