Gene Transcripts
- Gene Symbol
- Gm5512
- Entrez Gene ID
- 433224
- Gene Name
- required for meiotic nuclear division 1 pseudogene
- Gene Aliases
- EG433224
- Chromosome Location
- Chr. 19: 12905231 - 12906993 on Build GRCm38
- UniGene ID
- Mm.153111
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002891.1 |
|
1 |
1246 |
|
- Gene Symbol
- Rmnd1
- Entrez Gene ID
- 66084
- Gene Name
- required for meiotic nuclear division 1 homolog (S. cerevisiae)
- Gene Aliases
- 0610042C05Rik, AA408137, AI462664, AW536662
- Chromosome Location
- Chr. 10: 4403166 - 4432407 on Build GRCm38
- UniGene ID
- Mm.153111
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK205767.1 |
- |
4 |
295 |
|
AK215625.1 |
- |
4 |
274 |
|
BC019572.1 |
- |
8 |
583 |
|
View Full Product Details