Gene Transcripts

Gene Symbol
OSTCP1
Entrez Gene ID
202459
Gene Name
oligosaccharyltransferase complex subunit pseudogene 1
Gene Aliases
DC2L, OSTCL
Chromosome Location
Chr. 6: 158841117 - 158857632 on Build GRCh38
UniGene ID
Hs.487035
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028496.1 3 1058
GenBank mRNA BC024224.1 - 3 1062

View Full Product Details