Gene Transcripts
- Gene Symbol
- UMODL1-AS1
- Entrez Gene ID
- 150147
- Gene Name
- UMODL1 antisense RNA 1
- Gene Aliases
- C21orf128
- Chromosome Location
- Chr. 21: 42102134 - 42108534 on Build GRCh38
- UniGene ID
- Hs.558646
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027243.1 |
|
2 |
1836 |
|
GenBank mRNA |
AK090790.1 |
- |
2 |
1750 |
|
View Full Product Details