Gene Transcripts
- Gene Symbol
- LINC00479
- Entrez Gene ID
- 150135
- Gene Name
- long intergenic non-protein coding RNA 479
- Gene Aliases
- C21orf129, PRED76
- Chromosome Location
- Chr. 21: 41711520 - 41715775 on Build GRCh38
- UniGene ID
- Hs.350679
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027272.1 |
|
2 |
189 |
|
GenBank mRNA |
AK057397.1 |
- |
2 |
189 |
|
BC128198.1 |
- |
1 |
167 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB022346.1 |
- |
2 |
189 |
|
Chr. 21 - 41712981 - 41715775 [-] |
View Full Product Details