Gene Transcripts
- Gene Symbol
- PMCHL1
- Entrez Gene ID
- 5369
- Gene Name
- pro-melanin concentrating hormone like 1 (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 5: 22142352 - 22152272 on Build GRCh38
- UniGene ID
- Hs.247975
- Species
- Homo sapiens
- Gene Symbol
- PMCHL2
- Entrez Gene ID
- 5370
- Gene Name
- pro-melanin concentrating hormone like 2 (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 5: 71375785 - 71385993 on Build GRCh38
- UniGene ID
- Hs.742169
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY008412.1 |
- |
4 |
1706 |
|
EU921425.1 |
- |
5 |
501 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX091674.1 |
- |
4 |
374 |
|
Chr. 5 - 22142464 - 22152246 [+] |
View Full Product Details