Gene Transcripts
- Gene Symbol
- SCARNA15
- Entrez Gene ID
- 677778
- Gene Name
- small Cajal body-specific RNA 15
- Gene Aliases
- ACA45
- Chromosome Location
- Chr. 15: 82755945 - 82756071 on Build GRCh38
- UniGene ID
- Hs.743895
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003011.1 |
|
1 |
83 |
|
GenBank mRNA |
AJ609485.1 |
- |
1 |
83 |
|
- Gene Symbol
- SNHG21
- Entrez Gene ID
- 100505616
- Gene Name
- small nucleolar RNA host gene 21
- Gene Aliases
- FSD2-AS1, SCARNA15HG
- Chromosome Location
- Chr. 15: 82750564 - 82757206 on Build GRCh38
- UniGene ID
- Hs.513091
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL360137.1 |
- |
1 |
63 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX538250.1 |
- |
2 |
5195 |
|
Chr. 15 - 82750572 - 82757085 [+] |
View Full Product Details