Gene Transcripts

Gene Symbol
TTC39C
Entrez Gene ID
125488
Gene Name
tetratricopeptide repeat domain 39C
Gene Aliases
C18orf17, HsT2697
Chromosome Location
Chr. 18: 23992773 - 24135952 on Build GRCh38
UniGene ID
Hs.128576
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243425.1 NP_001230354.1 2 640
XM_005258198.4 XP_005258255.1 2 123
XM_017025550.1 XP_016881039.1 2 217
GenBank mRNA AK074875.1 - 2 262


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE465341.1 - 2 167 Chr. 18 - 24014893 - 24020288 [+]
GenBankmRNA BF197725.1 - 2 169 Chr. 18 - 24014892 - 24020183 [+]
GenBankmRNA AW612284.1 - 2 147 Chr. 18 - 24014913 - 24019987 [+]
GenBankmRNA BF477582.1 - 2 96 Chr. 18 - 24014964 - 24020255 [+]
GenBankmRNA AW138334.1 - 2 167 Chr. 18 - 24014893 - 24020012 [+]
GenBankmRNA AI681193.1 - 2 168 Chr. 18 - 24014892 - 24020218 [+]
GenBankmRNA AI523823.1 - 2 147 Chr. 18 - 24014913 - 24020227 [+]
GenBankmRNA AW470336.1 - 2 147 Chr. 18 - 24014916 - 24020076 [+]
GenBankmRNA AI632691.1 - 2 147 Chr. 18 - 24014913 - 24020080 [+]
GenBankmRNA AA972636.1 - 2 163 Chr. 18 - 24014892 - 24020152 [+]
GenBankmRNA BF058122.1 - 2 168 Chr. 18 - 24014892 - 24020182 [+]
GenBankmRNA AW139596.1 - 2 167 Chr. 18 - 24014893 - 24020231 [+]
GenBankmRNA W72622.1 - 164 Chr. - - [-]
GenBankmRNA AW206969.1 - 2 167 Chr. 18 - 24014893 - 24020112 [+]
GenBankmRNA AI985561.1 - 2 147 Chr. 18 - 24014913 - 24020198 [+]
GenBankmRNA AW149586.1 - 2 147 Chr. 18 - 24014913 - 24020225 [+]
GenBankmRNA AA806974.1 - 2 89 Chr. 18 - 24014971 - 24020221 [+]
GenBankmRNA AA987961.1 - 2 146 Chr. 18 - 24014909 - 24020231 [+]
GenBankmRNA AA947936.1 - 2 141 Chr. 18 - 24014917 - 24020222 [+]
GenBankmRNA AW015583.1 - 2 167 Chr. 18 - 24014893 - 24020197 [+]
GenBankmRNA BE327092.1 - 2 168 Chr. 18 - 24014892 - 24020266 [+]
GenBankmRNA AW015071.1 - 2 167 Chr. 18 - 24014893 - 24019973 [+]
GenBankmRNA BE673679.1 - 2 169 Chr. 18 - 24014892 - 24020254 [+]
GenBankmRNA AW138149.1 - 2 167 Chr. 18 - 24014893 - 24020105 [+]
GenBankmRNA BE326333.1 - 2 168 Chr. 18 - 24014892 - 24020195 [+]
GenBankmRNA AI701287.1 - 2 148 Chr. 18 - 24014916 - 24019993 [+]
GenBankmRNA BE219161.1 - 2 168 Chr. 18 - 24014892 - 24020226 [+]
GenBankmRNA BF057123.1 - 2 168 Chr. 18 - 24014892 - 24020196 [+]
GenBankmRNA BG149174.1 - 2 122 Chr. 18 - 24014938 - 24020213 [+]
GenBankmRNA AI334306.1 - 2 147 Chr. 18 - 24014913 - 24020211 [+]
GenBankmRNA AA826395.1 - 2 116 Chr. 18 - 24014943 - 24020118 [+]
GenBankmRNA BF108457.1 - 2 169 Chr. 18 - 24014892 - 24020141 [+]
GenBankmRNA AI696060.1 - 2 168 Chr. 18 - 24014892 - 24020187 [+]
GenBankmRNA HY051839.1 - 2 297 Chr. 18 - 24014763 - 24020000 [+]
GenBankmRNA AW052123.1 - 2 146 Chr. 18 - 24014914 - 24020192 [+]
GenBankmRNA AI298733.1 - 2 147 Chr. 18 - 24014913 - 24020203 [+]
GenBankmRNA AI767968.1 - 2 147 Chr. 18 - 24014913 - 24020218 [+]
GenBankmRNA AI475742.1 - 2 147 Chr. 18 - 24014913 - 24020202 [+]
GenBankmRNA AW167659.1 - 2 147 Chr. 18 - 24014913 - 24020177 [+]
GenBankmRNA AI350132.1 - 2 147 Chr. 18 - 24014913 - 24020166 [+]
GenBankmRNA BE467775.1 - 2 150 Chr. 18 - 24014910 - 24020083 [+]
GenBankmRNA AA488904.1 - 2 95 Chr. 18 - 24014968 - 24020108 [+]
GenBankmRNA BE328182.1 - 2 150 Chr. 18 - 24014910 - 24020079 [+]
GenBankmRNA BE674575.1 - 2 169 Chr. 18 - 24014892 - 24020195 [+]
GenBankmRNA AW016152.1 - 2 167 Chr. 18 - 24014893 - 24019911 [+]
GenBankmRNA AW968939.1 - 2 151 Chr. 18 - 24014909 - 24020259 [+]
GenBankmRNA AA410446.1 - 2 168 Chr. 18 - 24014892 - 24020160 [+]
GenBankmRNA HY026865.1 - 2 423 Chr. 18 - 24014637 - 24019941 [+]
GenBankmRNA BF433075.1 - 2 169 Chr. 18 - 24014892 - 24020285 [+]
GenBankmRNA BE217897.1 - 2 168 Chr. 18 - 24014892 - 24020255 [+]
GenBankmRNA AI801957.1 - 2 168 Chr. 18 - 24014892 - 24020229 [+]
GenBankmRNA BM701431.1 - 2 134 Chr. 18 - 24014926 - 24020187 [+]
GenBankmRNA BG150505.1 - 2 148 Chr. 18 - 24014912 - 24020058 [+]
GenBankmRNA DA734643.1 - 2 262 Chr. 18 - 24014799 - 24020073 [+]
GenBankmRNA BE222387.1 - 2 166 Chr. 18 - 24014894 - 24020083 [+]

View Full Product Details