Gene Transcripts
- Gene Symbol
- HSP90B3P
- Entrez Gene ID
- 343477
- Gene Name
- heat shock protein 90 beta family member 3, pseudogene
- Gene Aliases
- GRP94C, TRA1P2, TRAP2
- Chromosome Location
- Chr. 1: 91642518 - 91644341 on Build GRCh38
- UniGene ID
- Hs.591435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY956769.1 |
- |
3 |
1266 |
|
View Full Product Details