Gene Transcripts

Gene Symbol
Svet1
Entrez Gene ID
266615
Gene Name
subventricular expressed transcript 1
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.450551
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF323987.1 - LIT1755
Svet1
1 2219

View Full Product Details