Gene Transcripts
- Gene Symbol
- LOC101928269
- Entrez Gene ID
- 101928269
- Gene Name
- uncharacterized LOC101928269
- Gene Aliases
- Chromosome Location
- Chr. 21: 35954660 - 36004667 on Build GRCh38
- UniGene ID
- Hs.589909
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110418.1 |
|
4 |
515 |
|
GenBank mRNA |
BC032403.1 |
- |
2 |
288 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK123035.1 |
- |
|
249 |
|
Chr. - - [-] |
View Full Product Details