Gene Transcripts
- Gene Symbol
- LOC157503
- Entrez Gene ID
- 157503
- Gene Name
- uncharacterized LOC157503
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.103535
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055509.1 |
- |
1 |
1779 |
|
AK126867.1 |
- |
1 |
4474 |
|
View Full Product Details