Gene Transcripts
- Gene Symbol
- NTM-AS1
- Entrez Gene ID
- 101929637
- Gene Name
- NTM antisense RNA 1
- Gene Aliases
- C11orf39
- Chromosome Location
- Chr. 11: 131662129 - 131663556 on Build GRCh38
- UniGene ID
- Hs.730330
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127362.1 |
- |
1 |
1208 |
|
View Full Product Details