Gene Transcripts
- Gene Symbol
- HK2
- Entrez Gene ID
- 3099
- Gene Name
- hexokinase 2
- Gene Aliases
- HKII, HXK2
- Chromosome Location
- Chr. 2: 74832655 - 74893354 on Build GRCh38
- UniGene ID
- Hs.406266
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI085541.1 |
- |
1 |
418 |
|
BM912287.1 |
- |
|
91 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BC013295.1 |
- |
1 |
1591 |
|
Chr. 2 - 74833399 - 74835160 [-] |
View Full Product Details