Gene Transcripts
- Gene Symbol
- TBC1D27
- Entrez Gene ID
- 96597
- Gene Name
- TBC1 domain family member 27
- Gene Aliases
- Chromosome Location
- Chr. 17: 16922772 - 16926973 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024458.1 |
- |
7 |
2152 |
|
BC128223.1 |
- |
3 |
650 |
|
- Gene Symbol
- TBC1D26
- Entrez Gene ID
- 353149
- Gene Name
- TBC1 domain family member 26
- Gene Aliases
- Chromosome Location
- Chr. 17: 15732277 - 15744784 on Build GRCh38
- UniGene ID
- Hs.434123
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC028414.1 |
- |
5 |
905 |
|
View Full Product Details