Gene Transcripts

Gene Symbol
LOC101928907
Entrez Gene ID
101928907
Gene Name
uncharacterized LOC101928907
Gene Aliases
Chromosome Location
Chr. 15: 62060503 - 62062434 on Build GRCh38
UniGene ID
Hs.552949
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_135690.1 1 191
GenBank mRNA BC028842.1 - 1 163
BC034325.1 - 1 175
DA068710.1 - 1 191

View Full Product Details