Gene Transcripts
- Gene Symbol
- ZNF101
- Entrez Gene ID
- 94039
- Gene Name
- zinc finger protein 101
- Gene Aliases
- HZF12
- Chromosome Location
- Chr. 19: 19668153 - 19683506 on Build GRCh38
- UniGene ID
- Hs.631642
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU532885.1 |
- |
1 |
600 |
|
Chr. 19 - 19682676 - 19683398 [+] |
GenBankmRNA |
BX105661.1 |
- |
1 |
305 |
|
Chr. 19 - 19682973 - 19683509 [+] |
GenBankmRNA |
DB123786.1 |
- |
1 |
543 |
|
Chr. 19 - 19682730 - 19683331 [+] |
GenBankmRNA |
AA233033.1 |
- |
1 |
211 |
|
Chr. 19 - 19683081 - 19683305 [+] |
GenBankmRNA |
BF214783.1 |
- |
1 |
483 |
|
Chr. 19 - 19682783 - 19683479 [+] |
View Full Product Details