Gene Transcripts
- Gene Symbol
- PCOLCE-AS1
- Entrez Gene ID
- 100129845
- Gene Name
- PCOLCE antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 100589401 - 100604038 on Build GRCh38
- UniGene ID
- Hs.714123
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038910.1 |
|
7 |
1609 |
|
GenBank mRNA |
AK097705.1 |
- |
7 |
1609 |
|
AK298216.1 |
- |
8 |
1077 |
|
View Full Product Details