Gene Transcripts

Gene Symbol
QKI
Entrez Gene ID
9444
Gene Name
QKI, KH domain containing RNA binding
Gene Aliases
Hqk, QK, QK1, QK3, hqkI
Chromosome Location
Chr. 6: 163414486 - 163578596 on Build GRCh38
UniGene ID
Hs.510324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301085.1 NP_001288014.1 8 6539
NM_006775.2 NP_006766.1 8 6563
NM_206853.2 NP_996735.1 7 11236
NM_206854.2 NP_996736.1 7 12515
NM_206855.2 NP_996737.1 6 13462
XM_011536258.2 XP_011534560.1 8 6067
XM_011536260.2 XP_011534562.1 7 6461
GenBank mRNA AK096701.1 - 1 1453

View Full Product Details