Gene Transcripts
- Gene Symbol
- 4930426I24Rik
- Entrez Gene ID
- 100504512
- Gene Name
- RIKEN cDNA 4930426I24 gene
- Gene Aliases
- 6720480K08Rik
- Chromosome Location
- Chr. 12: 76389504 - 76403654 on Build GRCm38
- UniGene ID
- Mm.159844
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015206.1 |
- |
1 |
227 |
|
AK020166.1 |
- |
1 |
555 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CF198157.1 |
- |
1 |
150 |
|
Chr. 12 - 76402738 - 76403578 [-] |
View Full Product Details