Gene Transcripts

Gene Symbol
1700025M24Rik
Entrez Gene ID
72276
Gene Name
RIKEN cDNA 1700025M24 gene
Gene Aliases
Chromosome Location
Chr. 5: 73268580 - 73284184 on Build GRCm38
UniGene ID
Mm.271215
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040687.1 5 635
NR_040688.1 4 543
NR_040689.1 3 463
GenBank mRNA AK006359.1 - 3 466
BB014226.1 - 1 131
BC048655.1 - 5 632

View Full Product Details