Gene Transcripts
- Gene Symbol
- 2310061D13Rik
- Entrez Gene ID
- 69670
- Gene Name
- RIKEN cDNA 2310061D13 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK009996.1 |
- |
1 |
342 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY709562.1 |
- |
1 |
339 |
|
Chr. 12 - 79493258 - 79493635 [-] |
View Full Product Details