Gene Transcripts

Gene Symbol
Pdzd8
Entrez Gene ID
107368
Gene Name
PDZ domain containing 8
Gene Aliases
A630041P07Rik, AV329468, AW228944, Pdzk8
Chromosome Location
Chr. 19: 59296084 - 59345780 on Build GRCm38
UniGene ID
Mm.268797
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK088487.1 - 3294


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY762603.1 - 1 279 Chr. 19 - 59293231 - 59293890 [-]

View Full Product Details