Gene Transcripts
- Gene Symbol
- 4930435M08Rik
- Entrez Gene ID
- 74651
- Gene Name
- RIKEN cDNA 4930435M08 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.405588
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015326.1 |
- |
2 |
1132 |
|
View Full Product Details