Gene Transcripts
- Gene Symbol
- Ap2a1
- Entrez Gene ID
- 11771
- Gene Name
- adaptor-related protein complex 2, alpha 1 subunit
- Gene Aliases
- Adtaa
- Chromosome Location
- Chr. 7: 44900373 - 44929490 on Build GRCm38
- UniGene ID
- Mm.6877
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK078414.1 |
- |
1 |
696 |
|
View Full Product Details