Gene Transcripts
- Gene Symbol
- Gm35215
- Entrez Gene ID
- 102638721
- Gene Name
- predicted gene, 35215
- Gene Aliases
- Chromosome Location
- Chr. 13: 98936763 - 98945925 on Build GRCm38
- UniGene ID
- Mm.390635
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054100.1 |
- |
3 |
355 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB665859.1 |
- |
3 |
352 |
|
Chr. 13 - 98937797 - 98945915 [-] |
View Full Product Details