Gene Transcripts
- Gene Symbol
- B230117O15Rik
- Entrez Gene ID
- 78871
- Gene Name
- RIKEN cDNA B230117O15 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020978.1 |
- |
1 |
988 |
|
View Full Product Details